
Magnesium trifluoromethanesulphonate ≥98%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:60871-83-2
:
Formula:C₂F₆MgO₆S₂
MDL Number:MFCD00042601
:322.45 g/mol
Melting Pt:>310 °C
Storage Temperature:Ambient
UN:1759
ADR:8,III
:GHS05
Frequently Bought Together