Order Entry
Denmark
Orders LinkContactUsLinkComponent
Cyclohexane 99.5%, extra pure

Cyclohexane 99.5%, extra pure

Irritant
Health
Environment
Flammable
Cyclohexane 99.5%, extra pure
Certificates
Cyclohexane 99.5%, extra pure

3 Options Available Below

Product Details & Documents
Synonyms:  Hexahydrobenzene, Hexamethylene
  • CAS Number:110-82-7
  • Formula:C₆H₁₂
  • MDL Number:MFCD00003814
  • Molecular Weight:84.16 g/mol
  • Merck Index:13,02752
  • Boiling Pt:81 °C (760 mmHg)
  • Melting Pt:6.5 °C
  • Density:0.77 g/cm³
  • Flash Pt:–18 °C
  • Storage Temperature:Ambient
  • EINECS:203-806-2
  • UN:1145
  • ADR:3,II

Documents

Specifications for Product Family

Specification Test Results

  • Appearance:
    Clear liquid
  • Infrared spectrum:
    Conforms
  • Water:
    ≤0.01 %
  • GC:
    ≥99.5 %
  • Color scale:
    ≤10 APHA
  • Residue after evaporation:
    ≤0.005 %
Product Family Options

Product Information

  • Pack type
    Pk
    Availability
    Price
  • Glass bottle
    1 L
    Log in for availability
    Log in for availability
    1 * 1 L
    1 * 1 L - 658,00 kr
    658,00 kr
    1 * 1 L
       658,00 kr

  • Glass bottle
    2,5 L
    Log in for availability
    Log in for availability
    1 * 2,5 L
    1 * 2,5 L - 1.270,00 kr
    1.270,00 kr
    1 * 2,5 L
       1.270,00 kr

  • Plastic drum
    10 L
    Log in for availability
    Log in for availability
    1 * 10 L
    1 * 10 L - 3.180,00 kr
    3.180,00 kr
    1 * 10 L
       3.180,00 kr

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.