Organic reference standard, Nitrosodiphenylamine 1,000 µg/ml in dichloromethane
Catalog # CAMBULM-7219-1.2
Supplier: CAMBRIDGE ISOTOPE LABS
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:1,2 mL
- CAS No.:86-30-6
- Description:Nitrosodiphenylamine 1,000 µg/ml in dichloromethane
- Formula:C₁₂H₁₀N₂O
- Description:Nitrosodiphenylamine