Order Entry
Denmark
Orders LinkContactUsLinkComponent
Acenaphthene-[D₁₀] (99% D)

Acenaphthene-[D₁₀] (99% D)

Environment
Acenaphthene-[D₁₀] (99% D)
Certificates
Acenaphthene-[D₁₀] (99% D)

2 Options Available Below

Product Details & Documents
Synonyms:  Acenaphthene-[²H₁₀]
  • CAS Number:15067-26-2
  • Formula:C₁₂D₁₀
  • Molecular Weight:164.13 g/mol
  • Storage Temperature:Ambient
  • UN:3077
  • ADR:9,III
Product Family Options

Product Information

  • Pack type
    Pk
    Availability
    Price
  • Glass bottle for solids
    100 mg
    Log in for availability
    Log in for availability
    1 * 100 mg
    1 * 100 mg

  • Glass bottle for solids
    1 g
    Log in for availability
    Log in for availability
    1 * 1 g
    1 * 1 g

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.