Order Entry
Denmark
Orders LinkContactUsLinkComponent
Galactitol ≥97%
Galactitol ≥97%
Galactitol ≥97%
Galactitol ≥97%
Galactitol ≥97%
Purity: 97% CAS No.: 608-66-2 Molecular Formula: C6H14O6 Formula Weight: 182.17 Melting Point: 187-191° Boiling Point: 275-280°/1mm Merck Reference: 14,4332 MDL No.: MFCD00064288
 :  Dulcitol, rel-(2R,3S,4R,5S)-Hexane-1,2,3,4,5,6-hexaol
  •  :608-66-2
  •  :HOH₂C(CH(OH))₄CH₂OH
  •  :MFCD00064288
  •  :182.17 g/mol
  •  :14,4332
  •  :275…280 °C (1 mmHg)
  •  :187…191 °C
  •  :1.47
  •  :Ambient
  •  :210-165-2
Product Family Options
  • Pack type
    Pk
  • Glass bottle for solids
    25 g
    1 * 25 g
    1 * 25 g - 309,00 kr
    309,00 kr
    1 * 25 g
       309,00 kr
  • Glass bottle for solids
    100 g
    1 * 100 g
    1 * 100 g - 973,00 kr
    973,00 kr
    1 * 100 g
       973,00 kr
  • Glass bottle for solids
    500 g
    1 * 500 g
    1 * 500 g - 3.580,00 kr
    3.580,00 kr
    1 * 500 g
       3.580,00 kr