Ethylbenzene for synthesis, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
Synonyms: Phenylethane
CAS Number:100-41-4
:
Formula:C₈H₁₀
MDL Number:MFCD00011647
Boiling Pt:136.2
Melting Pt:–95 °C
Density:0.867 g/cm³ (20 °C)
Molecular Weight:106.17 g/mol
Flash Pt:15
EINECS:202-849-4
UN:1175
ADR:3 II
Merck Index:13,03801
:M-Clarity™ quality level = MQ200
:GHS02
:GHS08
:GHS07
Storage Temperature:Ambient
Specifications
- Environmentally Preferable:
Specifications
Specification Test Results
- Solubility:0.2 g/l (20 °C)
- Melting Point:-95 °C
- Molar Mass:106.16 g/mol
- Boiling Point:136 °C (1013 hPa)
- Vapor Pressure:9.5 hPa (20 °C)
- Flash Point:15 °C
- Explosion Limit:1.0 - 7.8 %(V)
- Density:0.87 g/cm3 (20 °C)
- Ignition Point:430 °C
Specification Test Results
Related Information
Frequently Bought Together