Tubing connectors, complete with screw cap, bent, DURAN®
SCOT286222005
- Thread:GL 14
- Tubing int.Ø:8 mm
- Pk:100
Screw cap PTB, red, with white PP bent hose connection.
- Very good chemical resistance
- Very good temperature resistance
With silicone seal (VMQ).