Order Entry
Denmark
Orders LinkContactUsLinkComponent
1-Chloro-2-phenylethane ≥99%

1-Chloro-2-phenylethane ≥99%

1-Chloro-2-phenylethane ≥99%
Certificates
1-Chloro-2-phenylethane ≥99%
1-Chloro-2-phenylethane ≥99%

3 Options Available Below

Product Details & Documents

Purity: 99% CAS No.: 622-24-2 Molecular Formula: C6H5(CH2)2Cl Formula Weight: 140.61 Flash Point: 66°(150°F) Refractive Index: 1.5300 Melting Point: -60° Boiling Point: 82-84°/16mm Beilstein Reference: 385969 MDL No.: MFCD00000977

Synonyms:  2-Phenylethyl chloride, (2-Chloroethyl)benzene, 2-(Chloroethyl)benzene
  • CAS Number:622-24-2
  • Formula:ClCH₂CH₂C₆H₅
  • MDL Number:MFCD00000977
  • Molecular Weight:140.61 g/mol
  • Boiling Pt:82…84 °C (16 mmHg)
  • Melting Pt:–60 °C
  • Density:1.070
  • Flash Pt:66 °C (150 °F)
  • Storage Temperature:Ambient
  • EINECS:210-725-6
Product Family Options

Product Information

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.