Spectrophotometers, GeneQuant™ 1300
Catalog #
Jump to:
Product Details & Documents
The GeneQuant 1300 spectrophotometer is pre-programmed with methods for nucleic acid purification, bacterial cell cultures, protein assays, Cy™Dye and cell culture density measurement.
- Other dyes into purified microarray hybridisation probes and PCR products
- Zoom facility, peak identification and on-peak confirmation
- Nucleic acid scanning to identify impurities, especially useful with RNA samples
cDNA application software for measuring incorporation of Cy™3 and Cy™5.
: USB cable and print via computer software supplied as standard.
Accessories
Product Information
- Catalog #AvailabilityPrice
- Spare papers for printer
Product Information
- Catalog #AvailabilityPrice
- Printer
Product Information
- Catalog #HeightVolumeAvailabilityPrice
- Rectangular cell with lid
Product Information
- Catalog #Path lengthWindow materialAvailabilityPrice
- Capillary cell for Genequant 100/300
Product Information
- Catalog #VolumePath lengthAvailabilityPrice
- Cell with stopper and black wallsCell with stopper and black wallsSCLI80-2103-6970 µl10 mmLog in for availability1 * 1 Items - 10.350,00 kr
Product Information
- Catalog #VolumePath lengthAvailabilityPrice
- Cell with stopper and black walls
Product Information
- Catalog #VolumePath lengthAvailabilityPrice
- Quartz cell