Order Entry
Denmark
Orders LinkContactUsLinkComponent
Stirrers, propeller, with two paddles
 :   BOCHEM
Stirrers, propeller, with two paddles
 :  BOCHEM
Stirrers, propeller, with two paddles
Stirrers, propeller, with two paddles
Propeller stirrer made of stainless steel with two flexible paddles.
  • Mixing of media with low and middle viscosity
  • Radial flow
Product Family Options
  • Description
    Stirrer Ø
    Shaft Ø
    Shaft length
  • Stirrer, with propeller and two flexible blades
    60 mm
    8 mm
    500 mm
    1 * 1 Items
    1 * 1 Items - 459,00 kr
    459,00 kr
    1 * 1 Items
       459,00 kr
  • Stirrer, with propeller and two flexible blades
    60 mm
    8 mm
    400 mm
    1 * 1 Items
    1 * 1 Items - 447,00 kr
    447,00 kr
    1 * 1 Items
       447,00 kr
  • Stirrer, with propeller and two flexible blades
    60 mm
    8 mm
    300 mm
    1 * 1 Items
    1 * 1 Items - 439,00 kr
    439,00 kr
    1 * 1 Items
       439,00 kr