Order Entry
Denmark
Orders LinkContactUsLinkComponent
Screwthread tubes, attachable, RD, DURAN®

Screwthread tubes, attachable, RD, DURAN®

RD 14, 14 mm, 1,5 mm
Catalog # SCOT286262206
Supplier:   DWK Life Sciences
Screwthread tubes, attachable, RD, DURAN®
Catalog # SCOT286262206
Supplier:  DWK Life Sciences
Screwthread tubes, attachable, RD, DURAN®
Screwthread tubes, attachable, RD, DURAN®

RD 14, 14 mm, 1,5 mm

Price
1.690,00 kr
Log in for availability
1 * 100 Items
Product Details & Documents

Made of DURAN®, borosilicate glass 3.3, clear.

  • Very good chemical resistance
  • High temperature resistance
  • Minimal thermal expansion giving relatively high resistance to temperature changes

Screw thread RD. It is used by glassblowers in the construction of laboratory glassware.

Specifications
  • Thread:
    RD 14
  • Tube ext.Ø:
    14 mm
  • Wall thickness:
    1,5 mm
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.