Screwthread tubes, GL, heavy wall DURAN®
Supplier: DWK Life Sciences
Jump to:
About this item
Made of DURAN®, borosilicate glass 3.3, clear.
- heavy wall
- Very good chemical resistance
- High temperature resistance
- Minimal thermal expansion giving relatively high resistance to temperature changes
Screw thread GL. It is used by glassblowers in the construction of laboratory glassware.
Supplementary to DIN 12216
5 Options Available Below
Product Details & Documents
Made of DURAN®, borosilicate glass 3.3, clear.
- heavy wall
- Very good chemical resistance
- High temperature resistance
- Minimal thermal expansion giving relatively high resistance to temperature changes
Screw thread GL. It is used by glassblowers in the construction of laboratory glassware.
Supplementary to DIN 12216
Product Family Options
Product Information
- ThreadTube ext.ØWall thicknessTotal lengthAvailabilityPrice