Ordreindtastning
Denmark
ContactUsLinkComponent
Target neddypningscylinder
Target neddypningscylinder
Katalog # BURK5365-6000
Supplier:  BURKLE
undefined
Target neddypningscylinder
Katalog # BURK5365-6000
Supplier:  BURKLE

Specifications

  • Ø:
    75 mm
  • Kapacitet:
    500 ml
  • Beskrivelse:
    Nedsænkningscylinder
  • Længde:
    210 mm
  • Pakn:
    1

Specifications

About this item

Precise liquid samples can be taken with this immersion cylinder. When the target has been reached, open the cover and pour out the sample.

  • Screw-off top enables easy cleaning
  • Air valve at the end of the tube
  • Venting lowering cable and cable reel