Order Entry
Denmark
ContactUsLinkComponent
Strips of 8 tubes
Strips of 8 tubes
Catalog # SIMPT110-15
Supplier:  Simport Scientific
undefined
Strips of 8 tubes
Catalog # SIMPT110-15
Supplier:  Simport Scientific
Specifications
  • Capacity:
    0,6 ml
  • Colour:
    Translucent
  • Description:
    Strips of 8 tubes
  • Material:
    PP
  • Pk:
    120
Specifications
About this item
Made of PP, translucent.
  • Very good chemical resistance
  • Very good temperature resistance
  • Certified RNase, DNase, Pyrogen and DNA-free
These plates feature 600 µl wells for smaller volume applications. They include a deep well plate along with twelve eight-tube strips. The tube strips are removable and can also be ordered separately.