Order Entry
Denmark
Orders LinkContactUsLinkComponent
Phenol, Supelco®

Phenol, Supelco®

Glass bottle for solids, 1 g
Catalog # 35952-1G
Toxic
Corrosive
Health
Supplier:   Merck
Phenol, Supelco®
Catalog # 35952-1G
Supplier:  Merck
CAS Number:  108-95-2
Phenol, Supelco®
Phenol, Supelco®

Glass bottle for solids, 1 g

CAS Number:  108-95-2
Price
620,00 kr
Log in for availability
1 * 1 g
Product Details & Documents
Synonyms:  Phenylalcohol, Carbolic acid, Hydroxybenzene
  • CAS Number:108-95-2
  • Formula:C₆H₅OH
  • MDL Number:MFCD00002143
  • Molecular Weight:94.11 g/mol
  • Merck Index:12,07390
  • Boiling Pt:181.7 °C (1013 hPa)
  • Melting Pt:40.8 °C
  • Density:1.0722 g/cm³ (20 °C)
  • Flash Pt:81 °C
  • UN:1671
  • ADR:6.1,II
  •  :M-Clarity™ quality level = MQ100
  • Storage Temperature:Ambient
  • EINECS:203-632-7
  • REACH:01-2119471329-32
Specifications

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    1 g

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.