Phenol, BAKER, J.T.Baker®
Plastic bottle for solids, 1 kg
Supplier: Avantor
Jump to:
Product Details & Documents
Synonyms: Phenylalcohol, Carbolic acid, Hydroxybenzene
- CAS Number:108-95-2
- Formula:C₆H₅OH
- MDL Number:MFCD00002143
- Molecular Weight:94.11 g/mol
- Merck Index:12,07390
- Boiling Pt:181.7 °C (1013 hPa)
- Melting Pt:40.8 °C
- Density:1.0722 g/cm³ (20 °C)
- Flash Pt:81 °C
- Storage Temperature:Ambient
- EINECS:203-632-7
- UN:1671
- ADR:6.1,II
- REACH:01-2119471329-32
Documents
Specifications
Specification Test Results
- Assay:99.0-100.5 %
- Acidity:passes test
- Appearance of solution:passes test
- Freezing Point:min. 39,5 °C
- Identification:passes test
- Residue on Evaporation:max. 0,05 %
Specifications
- Pack type:Plastic bottle for solids
- Pk:1 kg



