
1-Boc-4-piperidone 99%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:79099-07-3
:
Formula:C₁₀H₁₇NO₃
MDL Number:MFCD00151800
Melting Pt:74…75 °C
:199.25 g/mol
:GHS07
Storage Temperature:Ambient
Specification Test Results
- Appearance:White to light yellow Crystalline powder
- Infrared spectrum:Conforms
- Melting point:73 to 77 °C
- GC:≥98.5 %
Specification Test Results
Frequently Bought Together