Methylmagnesium bromide 12% (w/w) ∼1 mol/L in THF
Catalog # TCIAM0362-250G
Supplier: TCI
CAS Number:
Specifications
- Pk:250 g
- Seq:3545
Specifications
Specification Test Results
- Concentration (sec Butanol method):12.0 to 13.2 w/w%
Specification Test Results
About this item
:
:119.24 g/mol
EINECS:200-844-1
UN:3399
ADR:4.3,I
:GHS02
:GHS05
:GHS07
:GHS08
CAS Number:75-16-1
Formula:CH₃BrMg
MDL Number:MFCD00000041