4-Nitrophenyl phosphate, Sigma-Aldrich®
Catalog # 487663-10
Supplier: Merck Millipore (Oncogene)
CAS Number:
Specifications
- Pack type:Plastic bottle
- Pk:10 mL
- Seq:2275
Specifications
About this item
:
:219.09 g/mol
EINECS:206-353-9
CAS Number:330-13-2
Formula:C₆H₆NO₆P