

Lithium hexafluorophosphate ≥97.0% (by titrimetric analysis)
: TCI
:21324-40-3
:
:F₆LiP
:MFCD00011096
:~ 1,5 g/cm³ (20 °C)
:Ambient
:244-334-7
:2923
:8,II
:151.91 g/mol
:GHS05
:GHS06
- Appearance:White to Almost white powder to crystal
- Purity (Ion exchange titration):min. 97.0 %
Frequently Bought Together