Order Entry
Denmark
ContactUsLinkComponent
Ammonium peroxodisulphate (APS), powder
Oxidizing
Health
Irritant
Ammonium peroxodisulphate (APS), powder
Catalog # P011-B
Supplier:  G-Biosciences
CAS Number:  
undefined
Oxidizing
Health
Irritant
Ammonium peroxodisulphate (APS), powder
Catalog # P011-B
Supplier:  G-Biosciences
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    10 g
  • Seq:
    3236

Specifications

About this item

 :
 :228.2 g/mol
Merck Index:13,00545
Melting Pt:120 °C
Density:1.98 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:231-786-5
UN:1444
ADR:5.1,III
 :GHS03
 :GHS08
 :GHS07
CAS Number:7727-54-0
Formula:(NH₄)₂S₂O₈
MDL Number:MFCD00003390