Order Entry
Denmark
Orders LinkContactUsLinkComponent
Pharmacy scoops
Pharmacy scoops
Supplier:   BOCHEM
Pharmacy scoops
Supplier:  BOCHEM
Certificates
Pharmacy scoops
5 Options Available Below
Product Details & Documents
These chemical scoops made from stainless steel 18/10 are ideal for use in pharmaceutical and food industries.
  • With welded handle
  • Food contact approved
Product Family Options
Product Information
  • Capacity
    Length
    Ø
    Availability
    Price
  • 150 ml
    215 mm
    40 mm
    Log in for availability
    Log in for availability
    1 * 1 Items
    1 * 1 Items - 296,00 kr
    296,00 kr
    1 * 1 Items
       296,00 kr
  • 250 ml
    310 mm
    82 mm
    Log in for availability
    Log in for availability
    1 * 1 Items
    1 * 1 Items - 1.190,00 kr
    1.190,00 kr
    1 * 1 Items
       1.190,00 kr
  • 500 ml
    350 mm
    102 mm
    Log in for availability
    Log in for availability
    1 * 1 Items
    1 * 1 Items - 1.280,00 kr
    1.280,00 kr
    1 * 1 Items
       1.280,00 kr
  • 1000 ml
    390 mm
    120 mm
    Log in for availability
    Log in for availability
    1 * 1 Items
    1 * 1 Items - 1.430,00 kr
    1.430,00 kr
    1 * 1 Items
       1.430,00 kr
  • 2000 ml
    450 mm
    160 mm
    Log in for availability
    Log in for availability
    1 * 1 Items
    1 * 1 Items - 1.660,00 kr
    1.660,00 kr
    1 * 1 Items
       1.660,00 kr
Anbefalinger