Soil samplers, Puerkhauer
Supplier: BURKLE
Certificates
About this item
Very robust sampler for heavy and stony soils. The model with 1000 mm flute length has a conically-tapered 60 cm tip which makes it much easier to extract the driven-in sampler.
- Depth graduation of 10 cm divisions
- Attachable handle and non-slip plastic grips
- Reinforced impact head
Frequently Bought Together