Powder samplers, PharmaPicker
: BURKLE
PharmaPicker is designed for sampling very small quantities of highly potent or active powders for analysis. The smallest sample quantities can be collected with the PharmaPicker.
- Available separately or in a handy transport box
- Electro-polished stainless steel
- Can be extended repeatedly
: PharmaPicker case includes collection cylinders in both sizes, extension rods of 50 and 100 cm, a handle, volume tips (alternative sample volumes on request), and a transport box.
Frequently Bought Together