Order Entry
Denmark
ContactUsLinkComponent
2,4-Dinitroanisole
2,4-Dinitroanisole
Catalog # EHERC12782400
Supplier:  EHRENSTORFER
CAS Number:  
2,4-Dinitroanisole
Catalog # EHERC12782400
Supplier:  EHRENSTORFER
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    0,1 g
  • Seq:
    3020

Specifications

About this item

 :
Boiling Pt:351 °C (1013 hPa)
Boiling Pt:351 °C (1013 hPa)
Melting Pt:88 °C
Density:1.34 g/cm³ (20 °C)
 :198.14 g/mol
EINECS:204-310-9
 :GHS07
CAS Number:119-27-7
Formula:C₇H₆N₂O₅
MDL Number:MFCD00035745