Order Entry
Denmark
Orders LinkContactUsLinkComponent
Samplers, Liquid-Sampler
Samplers, Liquid-Sampler
Supplier:   BURKLE
Samplers, Liquid-Sampler
Supplier:  BURKLE
Certificates
Samplers, Liquid-Sampler
Samplers, Liquid-Sampler
4 Options Available Below
Product Details & Documents
Material: V4A stainless steel (1.4404). Sampler for thin to viscous liquids.
  • Easy one handed operation
  • Discharge funnel for safe filling of sample bottles
Documents
Product Family Options
Opened/closed with thumb
Product Information
  • Capacity
    Availability
    Price
  • 331-0032
    Liquid-Sampler, open with thumb, 32×460 mm
    Liquid-Sampler, open with thumb, 32×460 mm
    50 ml
    Log in for availability
    Log in for availability
    1 * 1 Items
    1 * 1 Items - 2.750,00 kr
    2.750,00 kr
    1 * 1 Items
       2.750,00 kr
  • 331-0033
    Liquid-Sampler, open with thumb, 32×540 mm
    Liquid-Sampler, open with thumb, 32×540 mm
    100 ml
    Log in for availability
    Log in for availability
    1 * 1 Items
    1 * 1 Items - 3.030,00 kr
    3.030,00 kr
    1 * 1 Items
       3.030,00 kr
Opened/closed semi-automatically
Product Information
  • Capacity
    Availability
    Price
  • 331-0035
    Liquid-Sampler, place onto bottle, 32×535 mm
    Liquid-Sampler, place onto bottle, 32×535 mm
    100 ml
    Log in for availability
    Log in for availability
    1 * 1 Items
    1 * 1 Items - 3.700,00 kr
    3.700,00 kr
    1 * 1 Items
       3.700,00 kr
  • 331-0034
    Liquid-Sampler, place onto bottle, 32×455 mm
    Liquid-Sampler, place onto bottle, 32×455 mm
    50 ml
    Log in for availability
    Log in for availability
    1 * 1 Items
    1 * 1 Items - 3.210,00 kr
    3.210,00 kr
    1 * 1 Items
       3.210,00 kr