Order Entry
Denmark
ContactUsLinkComponent
Beakers, low form, Azlon®
Beakers, low form, Azlon®
Catalog # 213-0098
Supplier:  SciLabware
Supplier #: BPB0250P
undefined
Beakers, low form, Azlon®
Catalog # 213-0098
Supplier:  SciLabware
Supplier #: BPB0250P
Log in for availability
Select an Option
80,5 mm
Select an Option
50 ml
Select an Option
250 ml
Select an Option
96 mm
Select an Option
04
Select an Option
10

About this item

PP, highly translucent.

  • Tapered stacking style; space saving and easy to handle
  • Good chemical resistance
  • Autoclavable at +121 °C

Ideal for general laboratory use. Crisp, blue, easy to read indelible graduations. Precise non drip pouring spouts.

Certifications: ISO 7056
Frequently Bought Together