Order Entry
Denmark
ContactUsLinkComponent
2,4-Dichlorophenol
2,4-Dichlorophenol
Catalog # EHERC12451000
Supplier:  EHRENSTORFER
CAS Number:  
2,4-Dichlorophenol
Catalog # EHERC12451000
Supplier:  EHRENSTORFER
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    0,25 g
  • Seq:
    3029

Specifications

About this item

 :
 :163 g/mol
Merck Index:13,03098
Boiling Pt:210 °C (1013 hPa)
Melting Pt:40…41 °C
Density:1.4 g/cm³ (20 °C)
Flash Pt:114 °C
EINECS:204-429-6
UN:2928
ADR:6.1,II
 :GHS06
 :GHS05
 :GHS09
Storage Temperature:Ambient
CAS Number:120-83-2
Formula:C₆H₄Cl₂O
MDL Number:MFCD00002169