Order Entry
Denmark
ContactUsLinkComponent
3,4-Dichloroanisole
3,4-Dichloroanisole
Catalog # EHERC12333400
Supplier:  EHRENSTORFER
CAS Number:  
3,4-Dichloroanisole
Catalog # EHERC12333400
Supplier:  EHRENSTORFER
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    0,1 g
  • Seq:
    3020

Specifications

About this item

 :
Boiling Pt:200…204 °C (1013 hPa)
Melting Pt:–7 °C
Density:1.33 g/cm³ (20 °C)
 :177.03 g/mol
CAS Number:36404-30-5
Formula:C₇H₆Cl₂O
MDL Number:MFCD00070785