Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 kr
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Denmark
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
4-Chloro-2-fluoroaniline
4-Chloro-2-fluoroaniline
4-Chloro-2-fluoroaniline
MOLE18590478-25G
:
Molekula
:
4-Chloro-2-fluoroaniline
MOLE18590478-25G
:
Molekula
:
Pack type:
Glass bottle for solids
Pk:
25 g
Seq:
3308
:
:
145.56 g/mol
:
104 °C (1013 hPa)
:
1.299 g/cm³ (20 °C)
:
261-034-1
:
2810
:
6.1,III
:
57946-56-2
:
C₆H₅ClFN
:
MFCD00010625