4-Chlorosalicylic acid 98%
Leverandør: Thermo Fisher Scientific
Certifikater
Om denne vare
CAS nummer:5106-98-9
:
Formel:C₇H₅ClO₃
MDL nummer:MFCD00002449
:172.57 g/mol
Smeltende Pt:209…213 °C
EINECS:225-829-7
:GHS07
Opbevaringstemperatur:Ambient
Specifikationstestresultater
- Appearance:Off-white to light beige Powder
- Infrared spectrum:Conforms
- Melting point:209 to 213 °C
- Titration with NaOH:≥97.5 %
Specifikationstestresultater
Frequently Bought Together