Order Entry
Denmark
ContactUsLinkComponent
2-Chlorobenzoic acid
Irritant
2-Chlorobenzoic acid
Catalog # EHERC11390500
Supplier:  EHRENSTORFER
CAS Number:  
Irritant
2-Chlorobenzoic acid
Catalog # EHERC11390500
Supplier:  EHRENSTORFER
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    0,25 g
  • Seq:
    3029

Specifications

About this item

 :
 :156.57 g/mol
Merck Index:13,02143
Boiling Pt:285 °C (1013 hPa)
Melting Pt:139…142 °C
Density:1.544 g/cm³ (20 °C)
Flash Pt:173 °C
Storage Temperature:Ambient
EINECS:204-285-4
 :GHS07
CAS Number:118-91-2
Formula:C₇H₅ClO₂
MDL Number:MFCD00002412