
2,3,4-Trihydroxybenzoic acid ≥97%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:610-02-6
:
Formula:C₇H₆O₅
MDL Number:MFCD00002447
:170.12 g/mol
Melting Pt:∼210 °C (dec.)
EINECS:210-205-9
:GHS07
Storage Temperature:Ambient
Related Information
Frequently Bought Together