Order Entry
Denmark
Orders LinkContactUsLinkComponent
Collection plates
Collection plates
Collection plate, PP, 96-well, V-bottom, V-bottom, 500 µl, 96
Catalog # 28-4039-43
Supplier:   Cytiva
Collection plates
Catalog # 28-4039-43
Supplier:  Cytiva
Collection plates
Collection plates
Collection plate, PP, 96-well, V-bottom, V-bottom, 500 µl, 96
Price
408,00 kr
Log in for availability
1 * 5 Items
Product Details & Documents
Made from biocompatible PP. Collection plates, 96-well, designed for collection of eluted samples in parallel from screening plates such as MultiTrap™ and PreDictor™ plates.
  • Convenient collection of samples in 96-well format
  • Excellent fit to prepacked MultiTrap plates and PreDictor plates
  • V-bottom minimises the loss of sample
Well volume: < 500 µl
Specifications
  • Form:
    V-bottom
  • No. of wells:
    96
  • Volume:
    500 µl
  • Description:
    Collection plate, PP, 96-well, V-bottom
  • Cat. No.:
    28-4039-43
Anbefalinger