Order Entry
Denmark
ContactUsLinkComponent
Microcentrifuge tubes, 1,5 ml
Microcentrifuge tubes, 1,5 ml
  KART279
 :  KARTELL
undefined
Microcentrifuge tubes, 1,5 ml
  KART279
 :  KARTELL
  • Type:
    Eppendorf type, with flat cap
  • Version:
    Graduated, conical bottom, non sterile
  • Capacity:
    1,5 ml
  • Pk:
    1.000
PP, translucent.
  • Very good chemical resistance
  • Very good temperature resistance
  • Flat cap with writing area
Ideal for samples analysis. Depending on centrifuge being used, these tubes can withstand a centrifugal force of up to 11000×g.