Bottle top tilt measures, without bottle
Catalog # HIRS300016060
Supplier: HIRSCHMANN
Specifications
- Capacity:10 ml
- Seq:02
- Pk:1
Specifications
About this item
Borosilicate glass 3,3.
- Very good chemical resistance
- High temperature resistance
- Minimal thermal expansion giving relatively high resistance to temperature changes
Automatic dispenser particularly suited for dispensing suspensions.
These tilt measures with ground joint cone NS 29/32 can fit on all bottles/flasks with NS29/32 sockets.
Ordering information: Reservoir bottles available on request.