Order Entry
Denmark
ContactUsLinkComponent
Deck kit for small bottle roller culture system, Wheaton®
Deck kit for small bottle roller culture system, Wheaton®
Catalog # 734-1170
Supplier:  DWK Life Sciences
undefined
Deck kit for small bottle roller culture system, Wheaton®
Catalog # 734-1170
Supplier:  DWK Life Sciences
Specifications
  • Description:
    Deck kit for small bottle roller culture system
  • Pk:
    1
Specifications
About this item
Attaches quickly and easily to the small bottle bench top roller culture system. Holds an additional two rollers per kit. These units will accept two additional decks. Kit consists of assembled roller deck, four support posts, drive belt, necessary hardware and assembly instructions.
Weight: 2,3 kg