Order Entry
Denmark
ContactUsLinkComponent
 
Sodium benzoate 99.0-100.5%, EMPROVE® ESSENTIAL Ph. Eur., BP, NF, FCC, E211, SAFC®
 
Sodium benzoate 99.0-100.5%, EMPROVE® ESSENTIAL Ph. Eur., BP, NF, FCC, E211, SAFC®
 

 

  • Assay (acidimetric, calc. on anhydrous substance):
    99.0 - 100.5 %
  • Assay (HPLC, calc. on anhydrous substance):
    99.0 - 101.0 %
  • Identity:
    passes test
  • Appearance of solution (100 g/l, water):
    passes test
  • Acidity or alkalinity:
    passes test
  • Chloride (Cl):
    ≤ 0.02 %
  • Sulphate (SO₄):
    ≤ 0.01 %
  • Total chlorine:
    ≤ 0.03 %
  • Heavy metals (as Pb):
    ≤ 10 ppm
  • As (Arsenic):
    ≤ 1 ppm
  • Cd (Cadmium):
    ≤ 10 ppm
  • Cu (Copper):
    ≤ 10 ppm
  • Hg (Mercury):
    ≤ 1 ppm
  • Pb (Lead):
    ≤ 2 ppm
  • Zn (Zinc):
    ≤ 10 ppm
  • Benzene:
    ≤ 2 ppm
  • Toluene:
    ≤ 890 ppm
  • Other residual solvents (ICH Q3C):
    excluded by production process
  • Multinuclear acids:
    passes test
  • Oxidizable matter:
    passes test
  • Water (according to Karl Fischer):
    ≤ 1.5 %

 

Frequently Bought Together