Gas washing bottles, with head, DURAN®
Supplier: DWK Life Sciences
Certificates
About this item
DURAN®, borosilicate glass 3.3, clear.
- Very good chemical resistance
- High temperature resistance
- Minimal thermal expansion giving relatively high resistance to temperature changes
- With fused-in filter disc
- With standard ground joint
Gas washing bottles, head with fritted disc and standard ground joint.
Frequently Bought Together