Order Entry
Denmark
Orders LinkContactUsLinkComponent
Vanillic acid ≥98%

Vanillic acid ≥98%

Vanillic acid ≥98%
Certificates
Vanillic acid ≥98%
Vanillic acid ≥98%

3 Options Available Below

Product Details & Documents

Purity: 98% CAS No.: 121-34-6 Molecular Formula: C8H8O4 Formula Weight: 168.15 Melting Point: 210-218° Beilstein Reference: 2208364 Merck Reference: 14,9931 MDL No.: MFCD00002551

Synonyms:  4-Hydroxy-3-methoxybenzoic acid
  • CAS Number:121-34-6
  • Formula:C₈H₈O₄
  • MDL Number:MFCD00002551
  • Molecular Weight:168.15 g/mol
  • Merck Index:14,9931
  • Boiling Pt:353 °C (1013 hPa)
  • Melting Pt:210…218 °C
  • Storage Temperature:Ambient
  • EINECS:204-466-8
Product Family Options

Product Information

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.