Aspirator bottles, narrow mouth, DURAN®
Supplier: DWK Life Sciences
Certificates
About this item
DURAN®, borosilicate glass 3.3, clear.
- Very good chemical resistance
- High thermal resistance, autoclavable
- Minimal thermal expansion giving relatively high resistance to temperature changes
- Glass Type I according to USP, EP and JP
Bottles, narrow mouth, ungrounded neck, with bottom tubulature.
With printed retrace code (batch identification).
Frequently Bought Together