Economy bottles, narrow neck, round, HDPE, environmental sample, with screw caps, Nalgene®
Supplier: Thermo Fisher Scientific
Certificates
About this item
HDPE, translucent with PP screw cap.
- Good chemical resistance
- Heat resistant from −100 to +120 °C
- Leakproof
Narrow neck economy bottles are lightweight versions of the original Nalgene® Lab Quality Bottle, suitable for lighter-duty, general purpose applications.
Specifications
- Catalog No:
- 331-4244
- 331-4243
- 331-4242
- 331-4241
- Environmentally Preferable:
- Low Manufacturing Impact, Product Transparency and Disclosure
- Low Manufacturing Impact, Product Transparency and Disclosure
- Low Manufacturing Impact, Product Transparency and Disclosure
- Low Manufacturing Impact, Product Transparency and Disclosure
- Environmental certification:
- NAR Certificate of Retirement, ACT Label
- NAR Certificate of Retirement, ACT Label
- NAR Certificate of Retirement, ACT Label
- IRECS; ISO 14001 (Certificate Registration No. EMS 752603), ACT Label