Order Entry
Denmark
ContactUsLinkComponent
1,2-Dihydroxybenzene-3,5-disulphonic acid disodium salt monohydrate indicator for complexometry, Fluka™
Irritant
1,2-Dihydroxybenzene-3,5-disulphonic acid disodium salt monohydrate indicator for complexometry, Fluka™
Certificates
Irritant
1,2-Dihydroxybenzene-3,5-disulphonic acid disodium salt monohydrate indicator for complexometry, Fluka™
Specification Test Results
  • Identity (IR):
    complying
  • Assay (HPLC):
    Min. 95.0 %
  • Indicator solution:
    complying
  • Water (Karl Fischer):
    Max. 7 %
  • Suitability for Complexometry:
    complying