Cholecalciferol (Vitamin D3)
Glass bottle for solids, 1 g
Catalog # SERA38340.01
Marketsource
Supplier: Serva
Jump to:
Product Details & Documents
Synonyms: Vitamin D3, Colecalciferol
- CAS Number:67-97-0
- Formula:C₂₇H₄₄O
- MDL Number:MFCD00078131
- Molecular Weight:384.65 g/mol
- EINECS:200-673-2
- UN:2811
- ADR:6.1,II
Specifications
Specifications
- Pack type:Glass bottle for solids
- Pk:1 g
Specifications
Anbefalinger
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



