Order Entry
Denmark
ContactUsLinkComponent
 
Pesticide reference standard, Pentachloroanisole 1000 µg/ml in methanol
 :  Restek
 
Pesticide reference standard, Pentachloroanisole 1000 µg/ml in methanol
 :  Restek

 

  • CAS No.:
    1825-21-4
  • Formula:
    C₇H₃Cl₅O
  • Description:
    Pentachloroanisole

 

Frequently Bought Together