Order Entry
Denmark
ContactUsLinkComponent
 
 
2-Ethylhexyl methacrylate stabilised, Sigma-Aldrich®
 :  Merck
 
 
2-Ethylhexyl methacrylate stabilised, Sigma-Aldrich®
 :  Merck

 

  • Solubility:
     - 0.1 g/l (20 °C)
  • Melting Point:
    -50 °C
  • Molar Mass:
    198.3 g/mol
  • Boiling Point:
    218 °C (1013 hPa)
  • Vapor Pressure:
    0.13 hPa (20 °C)
  • Flash Point:
    92 °C
  • Explosion Limit:
    0.6 %(V)
  • Density:
    0.88 g/cm3 (20 °C)
  • Ignition Point:
    380 °C

 

 

Frequently Bought Together