Order Entry
Denmark
ContactUsLinkComponent
 
 
2-Hydroxyethyl methacrylate ≥97.0% (by GC) stabilised for synthesis, Sigma-Aldrich®
 :  Merck
 
 
2-Hydroxyethyl methacrylate ≥97.0% (by GC) stabilised for synthesis, Sigma-Aldrich®
 :  Merck

 

  • Solubility:
    (20 °C) miscible
  • Melting Point:
    -60 °C
  • Molar Mass:
    130.14 g/mol
  • Boiling Point:
    250 °C (1013 hPa)
  • Vapor Pressure:
    1.3 hPa (20 °C)
  • Flash Point:
    101 °C
  • Refractive Index:
    1.4515 (20 °C, 589 nm)
  • Density:
    1.07 g/cm3 (20 °C)
  • Synonyms:
    2-Hydroxyethyl 2-methylpropenoate, HEMA, Methacrylic acid 2-hydroxyethyl ester
  • Hill Formula:
    C6H10O3
  • Chemical Formula:
    CH2=C(CH3)COOCH2CH2OH

 

 

Frequently Bought Together