Order Entry
Denmark
ContactUsLinkComponent
Organic reference standard, 3,4-Xylenol 100 µg/ml in methanol, Dr Ehrenstorfer
Organic reference standard, 3,4-Xylenol 100 µg/ml in methanol, Dr Ehrenstorfer
Catalog # EHERXA12734000ME
Supplier:  EHRENSTORFER
CAS Number:  
Organic reference standard, 3,4-Xylenol 100 µg/ml in methanol, Dr Ehrenstorfer
Catalog # EHERXA12734000ME
Supplier:  EHRENSTORFER
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    1 mL
  • CAS No.:
    95-65-8
  • Description:
    3,4-Xylenol 100 µg/ml in methanol
  • Formula:
    C₈H₁₀O
  • Description:
    3,4-Xylenol

Specifications

About this item