Cholecalciferol (Vitamin D3)
Catalog # EDQMC2100000
Supplier: EDQM
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:500 mg
- Seq:2725
Specifications
About this item
:
:384.65 g/mol
EINECS:200-673-2
UN:2811
ADR:6.1,II
CAS Number:67-97-0
Formula:C₂₇H₄₄O
MDL Number:MFCD00078131