Organic reference standard, 1,3-Dichloro-2-propanol 1000 µg/ml in methanol
Catalog # CAMBULM-8092-1.2
Supplier: CAMBRIDGE ISOTOPE LABS
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:1,2 mL
- CAS No.:96-23-1
- Description:1,3-Dichloro-2-propanol 1,000 µg/ml in methanol
- Formula:(ClCH₂)₂CHOH
- Description:1,3-Dichloro-2-propanol